Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TRMT9B
Entrez Gene ID
510845
Gene Name
tRNA methyltransferase 9B (putative)
Gene Aliases
C27H8ORF79, KIAA1456, TRM9L
Chromosome Location
Chr.27: 23989199 - 24059041 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076832.1 NP_001070300.1 3 - 4 553 69
GenBank mRNA BC123749.1 - 3 - 4 553 69

View Full Product Details