Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DZANK1
Entrez Gene ID
512107
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
C13H20ORF12
Chromosome Location
Chr.13: 38480518 - 38533139 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076838.1 NP_001070306.1 15 - 16 1646 127
GenBank mRNA BC123405.1 - 15 - 16 1646 127

View Full Product Details