Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRPSAP1
Entrez Gene ID
789409
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 55387190 - 55413082 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076857.2 NP_001070325.1 5 - 6 815 75
GenBank mRNA BC123542.1 - 5 - 6 816 75

View Full Product Details