Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS7BP
Entrez Gene ID
613327
Gene Name
regulator of G protein signaling 7 binding protein
Gene Aliases
-
Chromosome Location
Chr.20: 14840630 - 14973460 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076930.1 NP_001070398.1 5 - 6 1151 93
GenBank mRNA BC123745.1 - 5 - 6 1151 93

View Full Product Details