Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTSZ
Entrez Gene ID
404187
Gene Name
cathepsin Z
Gene Aliases
-
Chromosome Location
Chr.13: 57381213 - 57390707 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077835.1 NP_001071303.1 2 - 3 373 97
GenBank mRNA BC122603.1 - 2 - 3 373 97

View Full Product Details