Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAB3
Entrez Gene ID
788288
Gene Name
TGF-beta activated kinase 1 (MAP3K7) binding protein 3
Gene Aliases
MAP3K7IP3
Chromosome Location
Chr.X: 112675022 - 112703760 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080371.2 NP_001073840.2 2 - 3 244 100
GenBank mRNA BT026536.1 - 1 - 2 39 100

View Full Product Details