Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YWHAB
Entrez Gene ID
286863
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta
Gene Aliases
-
Chromosome Location
Chr.13: 73297853 - 73319790 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174794.2 NP_777219.2 3 - 4 629 103
GenBank mRNA AF043736.1 - 2 - 3 422 103
BC120112.1 - 3 - 4 629 103

View Full Product Details