Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AURKB
Entrez Gene ID
360192
Gene Name
aurora kinase B
Gene Aliases
AIM-1, ARK-2, STK-1, STK12
Chromosome Location
Chr.19: 27835751 - 27840897 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183084.2 NP_898907.2 7 - 8 986 77
GenBank mRNA BC123650.1 - 7 - 8 986 77

View Full Product Details