Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASL11B
Entrez Gene ID
528446
Gene Name
RAS like family 11 member B
Gene Aliases
-
Chromosome Location
Chr.6: 68530438 - 68534897 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015635.2 NP_001015635.1 3 - 4 464 126
GenBank mRNA BC142345.1 - 3 - 4 477 126
BT020927.1 - 3 - 4 396 126
BT029844.1 - 3 - 4 464 126

View Full Product Details