Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TKFC
Entrez Gene ID
512373
Gene Name
triokinase and FMN cyclase
Gene Aliases
DAK
Chromosome Location
Chr.29: 39842535 - 39854640 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024524.3 NP_001019695.2 5 - 6 877 98
GenBank mRNA BC126551.1 - 5 - 6 878 98
BT021593.1 - 5 - 6 884 98

View Full Product Details