Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
535122
Gene Name
transmembrane protein 268
Gene Aliases
C8H9orf91, C9orf91
Chromosome Location
Chr.8: 103800600 - 103825494 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024561.2 NP_001019732.1 2 - 3 287 123
GenBank mRNA BC133593.1 - 2 - 3 287 123
BT020721.1 - 1 - 2 193 123

View Full Product Details