Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
535122
Gene Name
transmembrane protein 268
Gene Aliases
C8H9orf91, C9orf91
Chromosome Location
Chr.8: 103800600 - 103825494 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024561.2 NP_001019732.1 3 - 4 395 71
GenBank mRNA BC133593.1 - 3 - 4 395 71
BT020721.1 - 2 - 3 301 71

View Full Product Details