Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCGB1D
Entrez Gene ID
338419
Gene Name
secretoglobin, family 1D, member 2
Gene Aliases
LppAB, SCGB1D2
Chromosome Location
Chr.29: 40681417 - 40684223 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077807.1 NP_001071275.1 2 - 3 297 130
GenBank mRNA BC126826.1 - 2 - 3 297 130

View Full Product Details