Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NHEDC1
Entrez Gene ID
509608
Gene Name
solute carrier family 9, subfamily B (cation proton antiporter 2), member 1
Gene Aliases
-
Chromosome Location
Chr.6: 21861360 - 21903266 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077892.2 NP_001071360.1 2 - 3 171 121
GenBank mRNA BC126670.1 - 2 - 3 172 121

View Full Product Details