Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RESP18
Entrez Gene ID
510410
Gene Name
regulated endocrine specific protein 18
Gene Aliases
-
Chromosome Location
Chr.2: 107270818 - 107276358 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077897.2 NP_001071365.1 4 - 5 444 81
GenBank mRNA BC126772.1 - 4 - 5 446 81

View Full Product Details