Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCHF3
Entrez Gene ID
520348
Gene Name
membrane associated ring-CH-type finger 3
Gene Aliases
MARCH3
Chromosome Location
Chr.7: 26947807 - 27104265 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077941.1 NP_001071409.1 1 - 2 245 102
GenBank mRNA BC126540.1 - 1 - 2 245 102

View Full Product Details