Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SASH3
Entrez Gene ID
534137
Gene Name
SAM and SH3 domain containing 3
Gene Aliases
HXCXORF9
Chromosome Location
Chr.X: 13811589 - 13827151 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077998.2 NP_001071466.1 5 - 6 723 65
GenBank mRNA BC126545.1 - 5 - 6 724 65

View Full Product Details