Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIX1
Entrez Gene ID
535033
Gene Name
limb and CNS expressed 1
Gene Aliases
-
Chromosome Location
Chr.7: 96536557 - 96592167 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078003.1 NP_001071471.1 2 - 3 382 60
GenBank mRNA BC126599.1 - 2 - 3 382 60

View Full Product Details