Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PELI1
Entrez Gene ID
535054
Gene Name
pellino E3 ubiquitin protein ligase 1
Gene Aliases
-
Chromosome Location
Chr.11: 62517235 - 62573790 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078004.1 NP_001071472.1 1 - 2 331 66
GenBank mRNA BC126542.1 - 1 - 2 331 66

View Full Product Details