Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY6H
Entrez Gene ID
618749
Gene Name
lymphocyte antigen 6 family member H
Gene Aliases
-
Chromosome Location
Chr.14: 1361015 - 1364698 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079636.2 NP_001073104.1 1 - 2 155 97
GenBank mRNA BC126596.1 - 1 - 2 156 97

View Full Product Details