Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL15
Entrez Gene ID
533987
Gene Name
methyltransferase like 15
Gene Aliases
METT5D1
Chromosome Location
Chr.15: 58806387 - 59076911 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079782.2 NP_001073250.1 2 - 3 388 79
GenBank mRNA BC126575.1 - 2 - 3 389 79

View Full Product Details