Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C4H7orf57
Entrez Gene ID
614195
Gene Name
chromosome 4 C7orf57 homolog
Gene Aliases
C7orf57
Chromosome Location
Chr.4: 7668402 - 7697286 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079785.1 NP_001073253.1 1 - 2 61 93
GenBank mRNA BC126755.1 - 1 - 2 61 93

View Full Product Details