Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDX2
Entrez Gene ID
505159
Gene Name
ferredoxin 2
Gene Aliases
FDX1L
Chromosome Location
Chr.7: 14841526 - 14846145 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080226.2 NP_001073695.1 4 - 5 421 74
GenBank mRNA BC122825.1 - 4 - 5 422 74

View Full Product Details