Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHBDD3
Entrez Gene ID
507674
Gene Name
rhomboid domain containing 3
Gene Aliases
-
Chromosome Location
Chr.17: 68547850 - 68554273 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080238.1 NP_001073707.1 3 - 4 1168 138
GenBank mRNA BC146065.1 - 6 - 7 1315 138
BT029783.1 - 3 - 4 1168 138

View Full Product Details