The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080240.1 | NP_001073709.1 | 1 - 2 | 286 | 119 |
GenBank mRNA | EF175214.1 | - | 1 - 2 | 286 | 119 |