Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMCH
Entrez Gene ID
508013
Gene Name
pro-melanin concentrating hormone
Gene Aliases
-
Chromosome Location
Chr.5: 66019749 - 66020914 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080240.1 NP_001073709.1 2 - 3 481 143
GenBank mRNA EF175214.1 - 2 - 3 481 143

View Full Product Details