Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOR3A
Entrez Gene ID
789634
Gene Name
torsin family 3 member A
Gene Aliases
-
Chromosome Location
Chr.16: 60422988 - 60450964 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080376.1 NP_001073845.1 3 - 4 689 67
GenBank mRNA BC126733.1 - 3 - 4 689 67

View Full Product Details