Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FXN
Entrez Gene ID
505694
Gene Name
frataxin
Gene Aliases
-
Chromosome Location
Chr.8: 45222629 - 45244976 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080727.1 NP_001074196.1 4 - 5 510 70
GenBank mRNA BC122620.1 - 4 - 5 510 70

View Full Product Details