Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPDEF
Entrez Gene ID
497620
Gene Name
SAM pointed domain containing ETS transcription factor
Gene Aliases
pdef
Chromosome Location
Chr.23: 8591536 - 8608900 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081578.1 NP_001075047.1 3 - 4 1062 79
GenBank mRNA AY862876.1 - 1 - 2 88 79
BC133366.1 - 3 - 4 1062 79

View Full Product Details