Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIFA
Entrez Gene ID
783855
Gene Name
TRAF interacting protein with forkhead associated domain
Gene Aliases
-
Chromosome Location
Chr.6: 13229159 - 13237465 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081621.1 NP_001075090.1 1 - 2 306 60
GenBank mRNA BC133302.1 - 1 - 2 306 60

View Full Product Details