Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC10A6
- Entrez Gene ID
-
540841
- Gene Name
- solute carrier family 10 (sodium/bile acid cotransporter), member 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 26441298 - 26469312 on Build ARS-UCD1.2
- Gene Symbol
- C6H4orf36
- Entrez Gene ID
-
616618
- Gene Name
- chromosome 6 C4orf36 homolog
- Gene Aliases
-
SLC10A6
- Chromosome Location
-
Chr.6: 101781495 - 101788941 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EF495206.1 |
- |
- |
692 |
69 |
- Gene Symbol
- LOC112441472
- Entrez Gene ID
-
112441472
- Gene Name
- solute carrier family 10 member 6-like
- Gene Aliases
-
SLC10A6
- Chromosome Location
-
Chr.6: 101728121 - 101747799 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EF495204.1 |
- |
- |
495 |
69 |
View Full Product Details