Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC10A6
Entrez Gene ID
540841
Gene Name
solute carrier family 10 (sodium/bile acid cotransporter), member 6
Gene Aliases
-
Chromosome Location
Chr.11: 26441298 - 26469312 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081738.1 NP_001075207.1 - 586 109
GenBank mRNA BC149199.1 - 2 - 3 645 109
DQ409211.1 - 3 - 4 229 109
EF186076.1 - - 586 109

View Full Product Details