Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTNAP3
Entrez Gene ID
784375
Gene Name
contactin associated protein-like 3
Gene Aliases
-
Chromosome Location
Chr.8: 65903969 - 66141830 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081745.1 NP_001075214.1 9 - 10 1618 61
GenBank mRNA BT029872.1 - 9 - 10 1618 61

View Full Product Details