The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082450.1 | NP_001075919.1 | 1 - 2 | 155 | 152 |
GenBank mRNA | BC133625.1 | - | 1 - 2 | 155 | 152 |