Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINS4
Entrez Gene ID
616799
Gene Name
GINS complex subunit 4
Gene Aliases
-
Chromosome Location
Chr.27: 36454833 - 36470326 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082460.2 NP_001075929.1 4 - 5 479 86
GenBank mRNA BC133560.1 - 4 - 5 480 86

View Full Product Details