Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNGR3
Entrez Gene ID
618415
Gene Name
synaptogyrin 3
Gene Aliases
-
Chromosome Location
Chr.25: 1544835 - 1549036 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082464.1 NP_001075933.1 1 - 2 230 67
GenBank mRNA BC133586.1 - 1 - 2 230 67

View Full Product Details