Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAG3
Entrez Gene ID
782633
Gene Name
BAG cochaperone 3
Gene Aliases
-
Chromosome Location
Chr.26: 39775458 - 39798895 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082471.2 NP_001075940.1 3 - 4 1089 134
GenBank mRNA BC133574.1 - 3 - 4 1090 134

View Full Product Details