Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIX1L
Entrez Gene ID
788858
Gene Name
limb and CNS expressed 1 like
Gene Aliases
-
Chromosome Location
Chr.3: 21418207 - 21443140 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082475.1 NP_001075944.1 3 - 4 669 117
GenBank mRNA BC133629.1 - 3 - 4 669 117

View Full Product Details