Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUSP23
Entrez Gene ID
513978
Gene Name
dual specificity phosphatase 23
Gene Aliases
-
Chromosome Location
Chr.3: 9912931 - 9914339 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082609.2 NP_001076078.1 1 - 2 27 60
GenBank mRNA BC133448.1 - 1 - 2 28 60
BC134731.1 - 1 - 2 38 60

View Full Product Details