Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF830
Entrez Gene ID
539497
Gene Name
zinc finger protein 830
Gene Aliases
CCDC16
Chromosome Location
Chr.19: 15118727 - 15120467 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082612.2 NP_001076081.1 1 - 1 1640 112
GenBank mRNA BC133450.1 - 1 - 1 1641 112

View Full Product Details