Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHL41
Entrez Gene ID
505794
Gene Name
kelch like family member 41
Gene Aliases
KBTBD10
Chromosome Location
Chr.2: 26734650 - 26750934 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083377.1 NP_001076846.1 2 - 3 1315 102
GenBank mRNA BC123835.1 - 2 - 3 1315 102

View Full Product Details