Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAIP1
Entrez Gene ID
510566
Gene Name
matrix AAA peptidase interacting protein 1
Gene Aliases
C2H2orf47, C2orf47
Chromosome Location
Chr.2: 88519446 - 88533145 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083406.2 NP_001076875.1 1 - 2 466 67
GenBank mRNA BC133463.1 - 1 - 2 467 67

View Full Product Details