Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICU2
Entrez Gene ID
512525
Gene Name
mitochondrial calcium uptake 2
Gene Aliases
EFHA1
Chromosome Location
Chr.12: 35458662 - 35512371 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083416.1 NP_001076885.1 9 - 10 930 136
GenBank mRNA BC123699.1 - 9 - 10 930 136

View Full Product Details