Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C20H5orf34
Entrez Gene ID
514072
Gene Name
chromosome 20 C5orf34 homolog
Gene Aliases
C5orf34
Chromosome Location
Chr.20: 31300239 - 31331079 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083426.1 NP_001076895.1 8 - 9 1509 133
GenBank mRNA BC114653.1 - 8 - 9 1509 133

View Full Product Details