Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKT1S1
Entrez Gene ID
514859
Gene Name
AKT1 substrate 1
Gene Aliases
-
Chromosome Location
Chr.18: 56216992 - 56225265 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083434.2 NP_001076903.1 1 - 2 401 128
GenBank mRNA BC133618.1 - 1 - 2 402 128

View Full Product Details