Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF19
Entrez Gene ID
532079
Gene Name
zinc finger protein 19
Gene Aliases
-
Chromosome Location
Chr.18: 39589643 - 39600605 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083460.2 NP_001076929.1 1 - 2 206 64
GenBank mRNA BC133549.1 - 1 - 2 207 64

View Full Product Details