Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIMS1
Entrez Gene ID
540281
Gene Name
LIM zinc finger domain containing 1
Gene Aliases
LIMS3, LIMS3L
Chromosome Location
Chr.11: 44713826 - 44796651 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083495.1 NP_001076964.1 6 - 7 730 73
GenBank mRNA BC123746.1 - 6 - 7 730 73

View Full Product Details