Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC7
Entrez Gene ID
613424
Gene Name
armadillo repeat containing 7
Gene Aliases
-
Chromosome Location
Chr.19: 56320651 - 56337275 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083504.1 NP_001076973.1 2 - 3 281 55
GenBank mRNA BC123844.1 - 2 - 3 281 55

View Full Product Details