Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAMP3
Entrez Gene ID
613547
Gene Name
receptor activity modifying protein 3
Gene Aliases
-
Chromosome Location
Chr.4: 76470713 - 76493525 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083505.2 NP_001076974.1 2 - 3 224 67
GenBank mRNA BC114856.1 - 2 - 3 225 67

View Full Product Details