Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOB4
Entrez Gene ID
781884
Gene Name
MOB family member 4, phocein
Gene Aliases
MOBKL3
Chromosome Location
Chr.2: 86060818 - 86090535 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083526.1 NP_001076995.1 6 - 7 471 94
GenBank mRNA BC114756.1 - 6 - 7 471 94

View Full Product Details